Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pga5
Entrez Gene ID
58803
Gene Name
pepsinogen 5, group I
Gene Aliases
1110035E17Rik, Pepf
Chromosome Location
Chr.19: 10668957 - 10678071 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021453.4 NP_067428.2 3 - 4 428 70
GenBank mRNA AF240776.1 - 3 - 4 337 70
AK004109.1 - 3 - 4 367 70
AK053965.1 - 3 - 4 429 70
AK168924.1 - 3 - 4 367 70
BC119523.2 - 3 - 4 386 70

View Full Product Details