The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021473.3 | NP_067448.1 | 3 - 4 | 544 | 73 |
GenBank mRNA | AF225564.1 | - | 3 - 4 | 449 | 73 |
AK005376.1 | - | 3 - 4 | 480 | 73 | |
AK009462.1 | - | 3 - 4 | 321 | 73 | |
AK011321.1 | - | 3 - 4 | 473 | 73 | |
AK011388.1 | - | 3 - 4 | 499 | 73 | |
AK011667.1 | - | 3 - 4 | 500 | 73 | |
AK011794.1 | - | 3 - 4 | 473 | 73 | |
AK011856.1 | - | 3 - 4 | 73 | ||
AK011906.1 | - | 3 - 4 | 478 | 73 | |
AK011908.1 | - | 3 - 4 | 513 | 73 | |
AK128906.1 | - | 3 - 4 | 266 | 73 | |
AK144198.1 | - | 3 - 4 | 500 | 73 | |
AK146081.1 | - | 3 - 4 | 498 | 73 | |
AK146278.1 | - | 3 - 4 | 500 | 73 | |
AK152086.1 | - | 3 - 4 | 479 | 73 | |
AK159573.1 | - | 3 - 4 | 498 | 73 | |
AK164461.1 | - | 3 - 4 | 499 | 73 | |
AK168950.1 | - | 3 - 4 | 495 | 73 | |
AK178751.1 | - | 1 - 2 | 79 | 73 | |
AK186333.1 | - | 3 - 4 | 301 | 73 | |
AK203039.1 | - | 1 - 2 | 79 | 73 | |
AY207463.1 | - | 3 - 4 | 521 | 73 | |
BC003850.1 | - | 3 - 4 | 328 | 73 | |
BC039926.1 | - | 3 - 4 | 493 | 73 | |
BC046762.1 | - | 3 - 4 | 450 | 73 |