The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021501.4 | NP_067476.2 | 9 - 10 | 1128 | 69 |
GenBank mRNA | AF109174.1 | - | 9 - 10 | 1330 | 69 |
AK131822.1 | - | - | 1131 | 69 | |
AK145367.1 | - | - | 1223 | 69 | |
AK147813.1 | - | 9 - 10 | 1159 | 69 | |
AK208285.1 | - | 3 - 4 | 344 | 69 | |
BC020476.1 | - | 8 - 9 | 1094 | 69 | |
BC025159.1 | - | 9 - 10 | 1128 | 69 |