The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159631.1 | NP_001153103.1 | 2 - 3 | 226 | 78 |
NM_021606.3 | NP_067619.1 | 2 - 3 | 219 | 78 | |
GenBank mRNA | AF218847.1 | - | 2 - 3 | 221 | 78 |
AK004925.1 | - | 2 - 3 | 196 | 78 | |
AK011519.1 | - | 2 - 3 | 205 | 78 | |
AK018168.1 | - | 2 - 3 | 209 | 78 | |
AK075717.1 | - | 2 - 3 | 193 | 78 | |
AK075836.1 | - | 2 - 3 | 205 | 78 | |
AK086700.1 | - | - | 224 | 78 | |
AK089919.1 | - | 2 - 3 | 180 | 78 | |
AK170757.1 | - | 2 - 3 | 226 | 78 | |
BC019524.1 | - | 2 - 3 | 288 | 78 | |
BY205493.1 | - | 2 - 3 | 226 | 78 |