Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nek6
Entrez Gene ID
59126
Gene Name
NIMA (never in mitosis gene a)-related expressed kinase 6
Gene Aliases
1300007C09Rik
Chromosome Location
Chr.2: 38511697 - 38587490 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159631.1 NP_001153103.1 8 - 9 852 109
NM_021606.3 NP_067619.1 8 - 9 845 109
GenBank mRNA AF218847.1 - 8 - 9 847 109
AK004925.1 - 8 - 9 109
AK011519.1 - 8 - 9 831 109
AK018168.1 - 8 - 9 834 109
AK029266.1 - 8 - 9 850 109
AK075717.1 - 8 - 9 819 109
AK075836.1 - 8 - 9 831 109
AK086700.1 - - 850 109
AK089919.1 - 8 - 9 806 109
AK170757.1 - 8 - 9 852 109
AK185671.1 - 1 - 2 113 109
BC019524.1 - 8 - 9 914 109

View Full Product Details