The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023324.2 | NP_075813.2 | 6 - 7 | 1116 | 64 |
GenBank mRNA | AF302503.1 | - | 5 - 6 | 1338 | 64 |
AK151660.1 | - | 6 - 7 | 1117 | 64 | |
AK152900.1 | - | 6 - 7 | 1117 | 64 | |
AK152997.1 | - | 6 - 7 | 1116 | 64 | |
AK153372.1 | - | 6 - 7 | 1117 | 64 | |
AK167525.1 | - | 6 - 7 | 1117 | 64 | |
BC016515.1 | - | 5 - 6 | 715 | 64 | |
BC026695.1 | - | 6 - 7 | 1080 | 64 |