Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fip1l1
Entrez Gene ID
66899
Gene Name
FIP1 like 1 (S. cerevisiae)
Gene Aliases
1300019H17Rik, Rje
Chromosome Location
Chr.5: 74535356 - 74598790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159573.1 NP_001153045.1 9 - 10 939 79
GenBank mRNA AK008561.1 - 9 - 10 911 79
AK168464.1 - 9 - 10 942 79
BC003263.1 - 10 - 11 966 79

View Full Product Details