The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159573.1 | NP_001153045.1 | 15 - 16 | 1441 | 59 |
NM_001159574.1 | NP_001153046.1 | 13 - 14 | 1306 | 59 | |
NM_024183.5 | NP_077145.2 | 14 - 15 | 1333 | 59 | |
GenBank mRNA | AK005061.1 | - | 14 - 15 | 1302 | 59 |
AK008561.1 | - | 15 - 16 | 1413 | 59 | |
AK049672.1 | - | 13 - 14 | 1292 | 59 | |
AK164105.1 | - | 14 - 15 | 1299 | 59 | |
AK168464.1 | - | 15 - 16 | 1445 | 59 | |
AK190620.1 | - | 1 - 2 | 47 | 59 | |
BC003263.1 | - | 15 - 16 | 1441 | 59 |