Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cuedc2
Entrez Gene ID
67116
Gene Name
CUE domain containing 2
Gene Aliases
3010002G01Rik
Chromosome Location
Chr.19: 46329812 - 46338660 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164290.1 NP_001157762.1 8 - 9 863 107
NM_001164291.1 NP_001157763.1 7 - 8 755 107
NM_001164292.1 NP_001157764.1 7 - 8 752 107
NM_001164293.1 NP_001157765.1 7 - 8 722 107
NM_001164294.1 NP_001157766.1 7 - 8 701 107
NM_001164295.1 NP_001157767.1 7 - 8 698 107
NM_024192.2 NP_077154.2 8 - 9 854 107
GenBank mRNA AB232359.1 - 6 - 7 653 107
AK003457.1 - 7 - 8 657 108
AK008842.1 - 7 - 8 718 107
AK013884.1 - 7 - 8 713 107
BC003442.1 - 8 - 9 818 107

View Full Product Details