The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025297.2 | NP_079573.2 | 1 - 2 | 204 | 71 |
GenBank mRNA | AK002317.1 | - | 1 - 2 | 203 | 71 |
AK002533.1 | - | 1 - 2 | 204 | 71 | |
BC003864.1 | - | 1 - 2 | 204 | 71 | |
BY017691.1 | - | 1 - 2 | 204 | 71 |