Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dtd1
Entrez Gene ID
66044
Gene Name
D-tyrosyl-tRNA deacylase 1
Gene Aliases
0610006H08Rik, DUE-B, Hars2
Chromosome Location
Chr.2: 144599953 - 144768747 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025314.3 NP_079590.1 5 - 6 742 64
GenBank mRNA AK002246.1 - 5 - 6 679 64
AK003594.1 - 5 - 6 714 64
AK011917.1 - 5 - 6 711 64
AK017565.1 - 5 - 6 666 64
AK158898.1 - 5 - 6 799 64
BC026537.1 - 5 - 6 715 64

View Full Product Details