The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025318.3 | NP_079594.1 | 1 - 2 | 107 | 137 |
GenBank mRNA | AK002385.1 | - | 1 - 2 | 93 | 136 |
AK003198.1 | - | 1 - 2 | 103 | 136 | |
AK004149.1 | - | 1 - 2 | 103 | 137 | |
AK012769.1 | - | 1 - 2 | 93 | 136 | |
BC022104.1 | - | 1 - 2 | 78 | 137 |