Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem42
Entrez Gene ID
66079
Gene Name
transmembrane protein 42
Gene Aliases
0610027O18Rik, 4933429E06Rik, AV003444, D9Ertd662e
Chromosome Location
Chr.9: 123021038 - 123023491 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025339.4 NP_079615.1 2 - 3 393 79
GenBank mRNA AK002688.1 - 2 - 3 369 79
AK012820.1 - 2 - 3 368 79
BC049583.1 - 2 - 3 393 79

View Full Product Details