The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025432.3 | NP_079708.2 | 2 - 3 | 274 | 74 |
GenBank mRNA | AK002658.1 | - | 3 - 4 | 345 | 74 |
AK004389.1 | - | - | 299 | 74 | |
AK007889.1 | - | 2 - 3 | 275 | 74 | |
AK008564.1 | - | 3 - 4 | 324 | 74 | |
AK077643.1 | - | - | 300 | 74 | |
AY318751.1 | - | - | 332 | 74 | |
BC034845.1 | - | - | 309 | 74 | |
BC061087.1 | - | - | 344 | 74 | |
BC099551.1 | - | - | 316 | 74 |