The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160145.1 | NP_001153617.1 | 4 - 5 | 502 | 80 |
NM_001160146.1 | NP_001153618.1 | 4 - 5 | 398 | 80 | |
NM_025439.3 | NP_079715.1 | 4 - 5 | 367 | 80 | |
GenBank mRNA | AK005248.1 | - | 4 - 5 | 354 | 80 |
AK021225.1 | - | 4 - 5 | 354 | 80 | |
AK134909.1 | - | 4 - 5 | 332 | 80 | |
AK154423.1 | - | 4 - 5 | 354 | 80 | |
AK218177.1 | - | 2 - 3 | 169 | 80 | |
BC016524.1 | - | 4 - 5 | 342 | 80 | |
BC020291.1 | - | 3 - 4 | 319 | 80 | |
BC048017.1 | - | 4 - 5 | 321 | 80 | |
BQ918002.1 | - | 4 - 5 | 589 | 80 | |
CA460035.1 | - | 4 - 5 | 404 | 80 | |
CX208138.1 | - | 4 - 5 | 398 | 80 |