The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082532.1 | NP_001076001.1 | 1 - 2 | 67 | 90 |
GenBank mRNA | AK007367.1 | - | 1 - 2 | 48 | 90 |
AK021110.1 | - | 1 - 2 | 48 | 90 | |
BU558514.1 | - | 1 - 2 | 70 | 90 |