The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160208.1 | NP_001153680.1 | 7 - 8 | 1320 | 63 |
NM_001160209.1 | NP_001153681.1 | 6 - 7 | 1407 | 63 | |
NM_025505.4 | NP_079781.2 | 8 - 9 | 1341 | 63 | |
GenBank mRNA | AK005489.1 | - | 6 - 7 | 1141 | 63 |
AK006544.1 | - | 7 - 8 | 1324 | 63 | |
AK029787.1 | - | 8 - 9 | 1342 | 63 | |
AK167493.1 | - | 6 - 7 | 1414 | 63 | |
AK168933.1 | - | 6 - 7 | 1168 | 63 | |
AK171167.1 | - | 6 - 7 | 1199 | 63 |