The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163684.1 | NP_001157156.1 | 8 - 9 | 865 | 53 |
NM_025533.3 | NP_079809.1 | 8 - 9 | 940 | 53 | |
GenBank mRNA | AK007962.1 | - | 8 - 9 | 814 | 53 |
AK010006.1 | - | 8 - 9 | 876 | 53 | |
AK076736.1 | - | 8 - 9 | 865 | 53 | |
BC089029.1 | - | 8 - 9 | 866 | 53 |