The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025539.2 | NP_079815.2 | 2 - 3 | 166 | 78 |
GenBank mRNA | AK009933.1 | - | 2 - 3 | 164 | 78 |
AK018771.1 | - | 2 - 3 | 164 | 78 | |
AK132532.1 | - | - | 167 | 78 | |
BC025153.1 | - | 1 - 2 | 142 | 78 |