Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tceanc2
Entrez Gene ID
66526
Gene Name
transcription elongation factor A (SII) N-terminal and central domain containing 2
Gene Aliases
2210010B22Rik, 2210012G02Rik, 6330404A07Rik, AU015593, Tdeanc2
Chromosome Location
Chr.4: 107134162 - 107178366 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025617.2 NP_079893.1 2 - 3 267 116
GenBank mRNA AK008715.1 - 2 - 3 222 116
AK011991.1 - 2 - 3 208 116
BC028321.1 - 2 - 3 205 116

View Full Product Details