The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025639.4 | NP_079915.1 | 1 - 2 | 120 | 69 |
NM_178269.3 | NP_840000.2 | 1 - 2 | 120 | 69 | |
GenBank mRNA | AK010885.1 | - | 1 - 2 | 112 | 69 |
AK011191.1 | - | 1 - 2 | 97 | 69 | |
AK011227.1 | - | 1 - 2 | 104 | 69 | |
AK011464.1 | - | 1 - 2 | 121 | 69 | |
AK011755.1 | - | 1 - 2 | 113 | 69 | |
AK011793.1 | - | 1 - 2 | 106 | 69 | |
AK017679.1 | - | 1 - 2 | 113 | 69 | |
AY243536.1 | - | 1 - 2 | 81 | 69 | |
AY243537.1 | - | 1 - 2 | 81 | 69 | |
BC009160.1 | - | 1 - 2 | 99 | 69 | |
BY710734.1 | - | 1 - 2 | 121 | 69 |