Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aste1
Entrez Gene ID
66595
Gene Name
asteroid homolog 1 (Drosophila)
Gene Aliases
1100001A21Rik, AV006403
Chromosome Location
Chr.9: 105395294 - 105408298 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025651.4 NP_079927.2 3 - 4 1801 59
GenBank mRNA AK003146.1 - 2 - 3 1138 59
AK030661.1 - 3 - 4 1672 59
BC048866.1 - 2 - 3 262 59
BC058377.1 - 3 - 4 1801 59
BC098471.1 - 4 - 5 1686 59

View Full Product Details