Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pspc1
Entrez Gene ID
66645
Gene Name
paraspeckle protein 1
Gene Aliases
5730470C09Rik, AI327109, AI449052, PSP1
Chromosome Location
Chr.14: 56722441 - 56778316 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025682.3 NP_079958.3 2 - 3 829 78
GenBank mRNA AK011272.1 - 2 - 3 826 78
AK017689.1 - 2 - 3 830 78
AK150823.1 - 2 - 3 830 78
AK160722.1 - 2 - 3 830 78
BC026772.1 - 2 - 3 808 78

View Full Product Details