Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d15
Entrez Gene ID
66687
Gene Name
TBC1 domain family, member 15
Gene Aliases
4432405K22Rik, AU022358, AU023730, AW557774, Ly6dl, Rab7-GAP
Chromosome Location
Chr.10: 115197871 - 115251493 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025706.3 NP_079982.3 3 - 4 293 86
GenBank mRNA AK010704.1 - 3 - 4 225 86
AK014477.1 - 3 - 4 225 86
AK078875.1 - 3 - 4 239 86
BB618866.1 - 3 - 4 297 86
BC032087.1 - 3 - 4 221 86
BC053395.1 - 3 - 4 274 86
DQ054831.1 - 3 - 4 224 86

View Full Product Details