Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nkapl
Entrez Gene ID
66707
Gene Name
NFKB activating protein-like
Gene Aliases
4921504I05Rik
Chromosome Location
Chr.13: 21467047 - 21468501 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025719.3 NP_079995.2 1 - 1 1265 118
GenBank mRNA AK014812.1 - 1 - 1 1252 118
AK160424.1 - 1 - 1 1067 118
BC099550.1 - 1 - 1 1265 118

View Full Product Details