The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168591.1 | NP_001162063.1 | 8 - 9 | 1172 | 89 |
NM_025827.3 | NP_080103.1 | 14 - 15 | 2476 | 89 | |
GenBank mRNA | AK003766.1 | - | 2 - 3 | 313 | 89 |
AK004843.1 | - | 14 - 15 | 2390 | 89 | |
AK050231.1 | - | 14 - 15 | 2385 | 89 | |
AK075719.1 | - | 8 - 9 | 1173 | 89 | |
AK088348.1 | - | 13 - 14 | 2382 | 89 | |
AK151103.1 | - | 14 - 15 | 2422 | 89 | |
AK151406.1 | - | 12 - 13 | 1990 | 89 | |
AK152964.1 | - | 12 - 13 | 1992 | 89 | |
AK168628.1 | - | 12 - 13 | 2000 | 89 | |
AK168791.1 | - | 14 - 15 | 2422 | 89 | |
AK168956.1 | - | 14 - 15 | 2422 | 89 | |
BC049090.1 | - | 14 - 15 | 2412 | 89 |