The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025901.3 | NP_080177.1 | 2 - 3 | 299 | 82 |
GenBank mRNA | AK005152.1 | - | 2 - 3 | 203 | 82 |
AK008637.1 | - | 2 - 3 | 171 | 82 | |
AK010875.1 | - | 2 - 3 | 237 | 82 | |
AK021353.1 | - | 2 - 3 | 234 | 82 | |
AK031110.1 | - | 2 - 3 | 234 | 82 | |
AK088206.1 | - | 2 - 3 | 199 | 82 | |
BC025898.1 | - | 2 - 3 | 315 | 82 |