The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025989.3 | NP_080265.2 | 3 - 4 | 671 | 80 |
GenBank mRNA | AK009323.1 | - | 3 - 4 | 672 | 80 |
AK009661.1 | - | 3 - 4 | 672 | 80 | |
AK009843.1 | - | 3 - 4 | 625 | 80 | |
BC119573.2 | - | 3 - 4 | 666 | 80 |