The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026002.4 | NP_080278.3 | 1 - 2 | 693 | 76 |
GenBank mRNA | AF501309.1 | - | 1 - 2 | 631 | 76 |
AK009253.1 | - | 1 - 2 | 607 | 76 | |
AK011808.1 | - | 1 - 2 | 688 | 76 | |
AK029915.1 | - | 1 - 2 | 694 | 76 | |
AK151269.1 | - | 1 - 2 | 677 | 76 | |
AK151810.1 | - | 1 - 2 | 688 | 76 | |
AY553638.1 | - | 1 - 2 | 376 | 76 | |
BC023115.1 | - | 1 - 2 | 592 | 76 | |
BC036994.1 | - | 1 - 2 | 412 | 76 | |
BC138858.1 | - | 1 - 2 | 407 | 76 | |
BC138859.1 | - | 1 - 2 | 407 | 76 |