The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026067.3 | NP_080343.4 | 5 - 6 | 844 | 94 |
GenBank mRNA | AK014041.1 | - | 5 - 6 | 845 | 94 |
AK014410.1 | - | 5 - 6 | 846 | 94 | |
AK144223.1 | - | 5 - 6 | 833 | 94 | |
AK165026.1 | - | 5 - 6 | 773 | 94 | |
AK172005.1 | - | 5 - 6 | 811 | 94 | |
BC046412.1 | - | 5 - 6 | 792 | 94 |