The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163767.1 | NP_001157239.1 | 4 - 5 | 542 | 112 |
NM_001163768.1 | NP_001157240.1 | 3 - 4 | 449 | 112 | |
NM_026100.3 | NP_080376.1 | 4 - 5 | 574 | 112 | |
GenBank mRNA | AK006804.1 | - | 4 - 5 | 543 | 112 |
AK015315.1 | - | 4 - 5 | 577 | 112 | |
BB634690.1 | - | 3 - 4 | 461 | 112 | |
BC048635.1 | - | 4 - 5 | 514 | 112 | |
BC099549.1 | - | 4 - 5 | 548 | 112 | |
BC132398.1 | - | 4 - 5 | 492 | 112 | |
BC132400.1 | - | 4 - 5 | 492 | 112 | |
BC145187.1 | - | 4 - 5 | 492 | 112 |