Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Armcx2
Entrez Gene ID
67416
Gene Name
armadillo repeat containing, X-linked 2
Gene Aliases
3230401N03Rik, AI043003, ALEX2
Chromosome Location
Chr.X: 134804142 - 134809221 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166397.1 NP_001159869.1 2 - 3 570 73
NM_001166398.1 NP_001159870.1 2 - 3 513 73
NM_026139.4 NP_080415.3 2 - 3 570 73
GenBank mRNA AK014329.1 - 2 - 3 346 73
AK034185.1 - 2 - 3 346 73
AK034621.1 - 2 - 3 344 73
AK043440.1 - 2 - 3 346 73
AK044926.1 - 2 - 3 344 73
AK053354.1 - 2 - 3 344 73
AK081299.1 - 2 - 3 289 73
AK172960.1 - 2 - 3 570 73
BC049088.1 - 2 - 3 325 73
BC052383.1 - 2 - 3 319 73

View Full Product Details