Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ciita
Entrez Gene ID
12265
Gene Name
class II transactivator
Gene Aliases
C2ta, EG669998, Gm9475, Mhc2ta
Chromosome Location
Chr.16: 10480015 - 10528606 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF042158.1 - 7 - 8 753 66
AF042159.1 - 8 - 9 1059 66
AF100709.1 - 7 - 8 936 66
AF100710.1 - 7 - 8 652 66
AK040723.1 - 7 - 8 815 66
AK138822.1 - 7 - 8 842 66
AK143956.1 - 7 - 8 924 66
AK153609.1 - 7 - 8 749 66
AK155184.1 - 7 - 8 776 66
AK155311.1 - 7 - 8 845 66
BC107330.1 - 7 - 8 694 66
BC107331.1 - 7 - 8 694 66
U60653.1 - 7 - 8 713 66

View Full Product Details