The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165902.1 | NP_001159374.1 | 3 - 4 | 333 | 62 |
NM_007614.3 | NP_031640.1 | 3 - 4 | 533 | 62 | |
GenBank mRNA | AK018515.1 | - | 3 - 4 | 447 | 62 |
AK020013.1 | - | 3 - 4 | 448 | 62 | |
AK035311.1 | - | 3 - 4 | 446 | 62 | |
AK133656.1 | - | 3 - 4 | 446 | 62 | |
AK199952.1 | - | 1 - 2 | 187 | 62 | |
AK201809.1 | - | 1 - 2 | 200 | 62 | |
AY751549.1 | - | 3 - 4 | 458 | 62 | |
BC048153.1 | - | 3 - 4 | 458 | 62 | |
BC053065.1 | - | 3 - 4 | 458 | 62 | |
M90364.1 | - | 3 - 4 | 343 | 62 |