The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007662.2 | NP_031688.2 | 1 - 2 | 155 | 78 |
GenBank mRNA | BC157978.1 | - | 1 - 2 | 72 | 78 |
M74541.1 | - | 1 - 2 | 562 | 78 |