Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cdh15
- Entrez Gene ID
-
12555
- Gene Name
- cadherin 15
- Gene Aliases
-
AI323380, Cdh14, Mcad
- Chromosome Location
-
Chr.8: 122848374 - 122867397 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M74541.1 |
- |
4 - 5 |
1019 |
80 |
View Full Product Details