Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdh16
Entrez Gene ID
12556
Gene Name
cadherin 16
Gene Aliases
-
Chromosome Location
Chr.8: 104601915 - 104624396 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252627.1 NP_001239556.1 5 - 6 766 78
NM_001252628.1 NP_001239557.1 6 - 7 856 78
NM_007663.3 NP_031689.1 6 - 7 856 78
GenBank mRNA AF016271.1 - 6 - 7 724 78
AJ609635.1 - 6 - 7 723 78
AK052636.1 - 6 - 7 659 78
AK093905.1 - 6 - 7 654 78
AK164546.1 - 5 - 6 767 78
BC015251.1 - 6 - 7 704 78

View Full Product Details