The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007694.4 | NP_031720.1 | 1 - 2 | 504 | 105 |
GenBank mRNA | AK019893.1 | - | 1 - 2 | 114 | 105 |
AK077487.1 | - | 1 - 2 | 70 | 105 | |
AK077489.1 | - | 1 - 2 | 112 | 105 | |
AK133629.1 | - | 1 - 2 | 112 | 105 | |
AK133739.1 | - | 1 - 2 | 112 | 105 | |
AK133786.1 | - | 1 - 2 | 114 | 105 | |
AK158857.1 | - | 1 - 2 | 505 | 105 | |
BC014736.1 | - | 1 - 2 | 105 | 105 | |
X51429.1 | - | 1 - 2 | 100 | 105 | |
X53028.1 | - | 1 - 2 | 110 | 105 |