The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007810.3 | NP_031836.1 | 8 - 9 | 1307 | 90 |
GenBank mRNA | AK143323.1 | - | 8 - 9 | 1387 | 90 |
BC103670.1 | - | 8 - 9 | 1290 | 90 | |
BC104489.1 | - | 8 - 9 | 1290 | 90 | |
D00659.1 | - | 8 - 9 | 1312 | 90 |