Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fer
Entrez Gene ID
14158
Gene Name
fer (fms/fps related) protein kinase
Gene Aliases
AV082135, C330004K01Rik, Fert, Fert2
Chromosome Location
Chr.17: 63829950 - 64146519 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008000.2 NP_032026.2 4 - 5 648 90
GenBank mRNA M32054.1 - 4 - 5 648 90

View Full Product Details