Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fli1
Entrez Gene ID
14247
Gene Name
Friend leukemia integration 1
Gene Aliases
EWSR2, Fli-1, SIC-1, Sic1
Chromosome Location
Chr.9: 32422204 - 32543427 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008026.5 NP_032052.1 6 - 7 968 79
GenBank mRNA AK036655.1 - 6 - 7 912 79
AK042111.1 - 6 - 7 881 79
AK083900.1 - 3 - 4 219 79
AK154714.1 - 6 - 7 885 79
BC138791.1 - 6 - 7 795 79
BC138792.1 - 6 - 7 795 79
X59421.1 - 6 - 7 968 79

View Full Product Details