The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077411.2 | NP_001070879.1 | 10 - 11 | 1613 | 68 |
NM_008094.5 | NP_032120.1 | 11 - 12 | 1782 | 68 | |
NR_122037.1 | 11 - 12 | 1818 | 68 | ||
GenBank mRNA | AK082767.1 | - | 10 - 11 | 1611 | 68 |
AK152745.1 | - | 10 - 11 | 1594 | 68 | |
AK166198.1 | - | 11 - 12 | 1810 | 68 | |
BC006663.1 | - | 11 - 12 | 1767 | 68 | |
M24119.1 | - | 10 - 11 | 1522 | 68 |