Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Man1a
Entrez Gene ID
17155
Gene Name
mannosidase 1, alpha
Gene Aliases
Man1a1, PCR1
Chromosome Location
Chr.10: 53904785 - 54076586 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008548.4 NP_032574.1 2 - 3 1126 77
GenBank mRNA AK028996.1 - 2 - 3 1041 77
AK031038.1 - 2 - 3 1127 77
AK042371.1 - 2 - 3 1062 77
AK043932.1 - 2 - 3 1067 77
AK148407.1 - 2 - 3 1067 77
AK154820.1 - 2 - 3 1068 77
AK171841.1 - 2 - 3 1069 77
AK172376.1 - 2 - 3 1069 77
AK172667.1 - 2 - 3 1068 77
BC015265.1 - 2 - 3 1027 77
U04299.1 - 1 - 2 730 77

View Full Product Details