Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrc1
Entrez Gene ID
17533
Gene Name
mannose receptor, C type 1
Gene Aliases
AW259686, CD206, MR
Chromosome Location
Chr.2: 14229414 - 14332023 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008625.2 NP_032651.2 11 - 12 1880 73
GenBank mRNA BC141338.1 - 11 - 12 1832 73
Z11974.1 - 11 - 12 1880 73

View Full Product Details