Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtf1
Entrez Gene ID
17764
Gene Name
metal response element binding transcription factor 1
Gene Aliases
MTF-1, Thyls
Chromosome Location
Chr.4: 124802549 - 124849800 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008636.4 NP_032662.3 5 - 6 999 81
GenBank mRNA AK150295.1 - 6 - 7 1061 81
AY260849.1 - 4 - 5 853 81
AY260850.1 - 4 - 5 853 81
BC017679.1 - 5 - 6 961 81
X71327.1 - 5 - 6 949 81

View Full Product Details