The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009008.3 | NP_033034.1 | 5 - 6 | 587 | 86 |
GenBank mRNA | AK007561.1 | - | 5 - 6 | 525 | 86 |
AK088415.1 | - | 5 - 6 | 536 | 86 | |
AK088805.1 | - | 5 - 6 | 524 | 86 | |
AK144136.1 | - | 5 - 6 | 531 | 86 | |
AK171321.1 | - | 5 - 6 | 588 | 86 | |
BC005455.1 | - | 5 - 6 | 503 | 86 | |
X53247.1 | - | 5 - 6 | 485 | 86 |