The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122958.1 | NP_001116430.1 | 18 - 19 | 2322 | 58 |
NM_001122959.1 | NP_001116431.1 | 18 - 19 | 2301 | 58 | |
NM_009015.3 | NP_033041.3 | 17 - 18 | 2652 | 58 | |
GenBank mRNA | AK030753.1 | - | 18 - 19 | 2280 | 58 |
AK088876.1 | - | 18 - 19 | 2319 | 58 | |
AK161644.1 | - | 18 - 19 | 2275 | 58 | |
BC021643.1 | - | 17 - 18 | 2633 | 58 | |
X97796.1 | - | 17 - 18 | 2112 | 58 |