Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rptn
Entrez Gene ID
20129
Gene Name
repetin
Gene Aliases
-
Chromosome Location
Chr.3: 93393699 - 93399442 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009100.2 NP_033126.2 1 - 2 47 63

View Full Product Details