Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Satb1
Entrez Gene ID
20230
Gene Name
special AT-rich sequence binding protein 1
Gene Aliases
2610306G12Rik, AW413156
Chromosome Location
Chr.17: 51736187 - 51833497 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163630.1 NP_001157102.1 6 - 7 1070 104
NM_001163631.1 NP_001157103.1 6 - 7 938 104
NM_001163632.1 NP_001157104.1 6 - 7 911 104
NM_009122.2 NP_033148.2 6 - 7 961 104
GenBank mRNA AK037740.1 - 6 - 7 961 104
AK088563.1 - 6 - 7 937 104
AK134040.1 - 6 - 7 941 104
AK158518.1 - 6 - 7 1071 104
BC011132.1 - 6 - 7 821 104
CT010371.1 - 5 - 6 749 104
U05252.1 - 6 - 7 947 104

View Full Product Details