Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sgca
Entrez Gene ID
20391
Gene Name
sarcoglycan, alpha (dystrophin-associated glycoprotein)
Gene Aliases
50DAG, Asg
Chromosome Location
Chr.11: 94962777 - 94976327 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009161.4 NP_033187.1 9 - 10 1241 82
GenBank mRNA AB024920.1 - 9 - 10 1241 82
AF019564.1 - 8 - 9 988 82
AK009015.1 - 8 - 9 999 82
AK075915.1 - 8 - 9 1013 82
BC060287.1 - 8 - 9 993 82

View Full Product Details